Triethylene glycol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol
95126-100ML-F 95126-500ML-F
SIAL95126-100ML-FEA 44.7 EUR
SIAL95126-100ML-F SIAL95126-500ML-F
Triethylene glycol, Sigma-Aldrich®
Triethylene glycol
Formula: C₆H₁₄O₄
MW: 150.17 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: –7 °C
Density: 1.125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002880
CAS Number: 112-27-6
EINECS: 203-953-2
Merck Index: 12,09802