(±)-1,2-Propanediol, Sigma-Aldrich®

Supplier: Merck

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

M-Clarity™ quality level = MQ200

P4347-4L P4347-500ML
SIALP4347-4LEA 1060 EUR
SIALP4347-4L SIALP4347-500ML
(±)-1,2-Propanediol, Sigma-Aldrich®
Propylene glycol
Formula: CH₃CH(OH)CH₂OH
Boiling Pt: 187.6 °C (1013 hPa)
Melting Pt: –59 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR