2,6-Bis(hydroxymethyl)-p-cresol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Methyl-2,6-bis(hydroxymethyl)phenol, 1,3-Bis(hydroxymethyl)-2-hydroxy-5-methylbenzene, 2,6-Bis(hydroxymethyl)-4-methylphenol, 3,5-Bis(hydroxymethyl)-4-hydroxytoluene
M-Clarity™ quality level = MQ100
227528-100G 227528-250G 227528-25G
SIAL227528-100GEA 157 EUR
SIAL227528-100G SIAL227528-250G SIAL227528-25G
2,6-Bis(hydroxymethyl)-p-cresol, Sigma-Aldrich®
2,6-Bis(hydroxymethyl)-p-cresol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 133…134 °C
Density: 1.382 g/cm³ (–173 °C)
MDL Number: MFCD00004619
CAS Number: 91-04-3
EINECS: 202-036-4