4-Nitro-2,6-xylenol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Hydroxy-5-nitro-3-xylene, 2,6-Dimethyl-4-nitrophenol

132713-10G 132713-1G
SIAL132713-10GEA 112 EUR
SIAL132713-10G SIAL132713-1G
4-Nitro-2,6-xylenol, Sigma-Aldrich®
4-Nitro-2,6-xylenol
Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00007339
CAS Number: 2423-71-4
EINECS: 219-353-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR