6-Nitro-2,4-xylenol ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: 2,4-Dimethyl-6-nitrophenol, 4-Hydroxy-5-nitro-3-xylene
B25205.06 B25205.14 B25205.22
B25205.06EA 46.7 EUR
B25205.06 B25205.14 B25205.22
6-Nitro-2,4-xylenol ≥98%
6-Nitro-2,4-xylenol
Purity: 98% CAS No.: 14452-34-7 Molecular Formula: C8H9NO3 Formula Weight: 167.17 Melting Point: 68-71° MDL No.: MFCD00191633
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00191633
CAS Number: 14452-34-7
EINECS: 238-436-0