1,4-Phenyldiboronic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
p-Phenylenebisboronic acid, 1,4-Phenylenediboronic acid, 1,4-Phenyldiboric acid, 1,4-Benzenediboronic acid
M-Clarity™ quality level = MQ100
417130-25G
417130-5G
SIAL417130-25GEA
257
EUR
SIAL417130-25G
SIAL417130-5G
1,4-Phenyldiboronic acid, Sigma-Aldrich®
1,4-Phenyldiboronic acid
Formula:
C₆H₈B₂O₄ MW: 165.75 g/mol |
MDL Number:
MFCD00236018 CAS Number: 4612-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...