1,5-Diamino-2-methylpentane, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Methyl-1,5-diaminopentane, 2-Methylpentane-1,5-diamine, 1,5-Diamino-2-methylpentane, 2-Methyl-1,5-pentanediamine
M-Clarity™ quality level = MQ200
329665-25ML 329665-1L
SIAL329665-25MLEA 67.5 EUR
SIAL329665-25ML SIAL329665-1L
1,5-Diamino-2-methylpentane, Sigma-Aldrich®
1,5-Diamino-2-methylpentane
Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 188…190 °C (1013 hPa)
Melting Pt: –60…–50 °C
Density: 0.86 g/cm³ (25 °C)
Flash Pt: 80 °C
MDL Number: MFCD00013460
CAS Number: 15520-10-2
EINECS: 239-556-6