16-Hydroxypalmitic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
16-Hydroxyhexadecylic acid, 16-Hydroxyhexadecanoic acid
M-Clarity™ quality level = MQ100
177490-1G
177490-5G
SIAL177490-1GEA
88.2
EUR
SIAL177490-1G
SIAL177490-5G
16-Hydroxypalmitic acid, Sigma-Aldrich®
16-Hydroxypalmitic acid
Formula:
C₁₆H₃₂O₃ MW: 272.43 g/mol Boiling Pt: 402 °C (1013 hPa) Melting Pt: 95 °C |
MDL Number:
MFCD00002750 CAS Number: 506-13-8 EINECS: 208-028-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...