N,N-Diethylethylenediamine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-(Diethylamino)ethylamine, 2-Aminoethyldiethylamine
M-Clarity™ quality level = MQ100
112720-100G 112720-5G
SIAL112720-100GEA 98.1 EUR
SIAL112720-100G SIAL112720-5G
N,N-Diethylethylenediamine, Sigma-Aldrich®
N,N-Diethylethylenediamine
Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 36 °C
MDL Number: MFCD00008176
CAS Number: 100-36-7
EINECS: 202-844-7
UN: 2685
ADR: 8,II