2-Amino-p-cresol 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3-Amino-4-hydroxytoluene, 1-Amino-2-hydroxy-5-methylbenzene, 2-Amino-4-methylphenol, 2-Hydroxy-5-methylaniline
163212500 163210500
ACRO163212500EA 67.7 EUR
ACRO163212500 ACRO163210500
2-Amino-p-cresol 97%
2-Amino-p-cresol
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259
Melting Pt: 134…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00007699
CAS Number: 95-84-1
EINECS: 202-457-3
Order Now
Specification Test Results
Appearance Beige to grey-brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 134 to 139 °C
GC ≥96.0 %