5-Amino-2-methylphenol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5-Amino-o-cresol, 3-Hydroxy-4-methylaniline, 1-Amino-3-hydroxy-4-methylbenzene, 5-Amino-2-methylphenol, 4-Amino-2-hydroxytoluene
M-Clarity™ quality level = MQ100
225088-25G
225088-5G
SIAL225088-25GEA
73.4
EUR
SIAL225088-25G
SIAL225088-5G
5-Amino-2-methylphenol, Sigma-Aldrich®
5-Amino-2-methylphenol
Formula:
C₇H₉NO MW: 123.15 g/mol Boiling Pt: 259 °C (1013 hPa) Melting Pt: 162 °C |
MDL Number:
MFCD00043922 CAS Number: 2835-95-2 EINECS: 220-618-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...