4-Amino-m-cresol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Amino-5-hydroxytoluene, 4-Amino-3-methylphenol, 1-Amino-4-hydroxy-2-methylbenzene, 4-Hydroxy-2-methylaniline

M-Clarity™ quality level = MQ200

144894-25G 144894-5G
SIAL144894-25GEA 137 EUR
SIAL144894-25G SIAL144894-5G
4-Amino-m-cresol, Sigma-Aldrich®
4-Amino-m-cresol
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 259
Melting Pt: 177…179 °C
MDL Number: MFCD00007871
CAS Number: 2835-99-6
EINECS: 220-621-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR