4-Chloro-3,5-xylenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Chloro-5-hydroxy-3-xylene, Chloroxylenol, 4-Chloro-3,5-dimethylphenol
M-Clarity™ quality level = MQ200
C38303-100G C38303-500G
SIALC38303-100GEA 38.1 EUR
SIALC38303-100G SIALC38303-500G
4-Chloro-3,5-xylenol, Sigma-Aldrich®
4-Chloro-3,5-xylenol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 114…116 °C
Density: 1.364 g/cm³ (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00002324
CAS Number: 88-04-0
EINECS: 201-793-8
Merck Index: 13,02194