5-Isopropyl-2-methylphenol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Carvacrol, 5-Isopropyl-o-cresol, Isothymol, 5-Isopropyl-2-methylphenol
M-Clarity™ quality level = MQ300
W224502-100G-K W224502-1KG-K W224502-5KG-K
SIALW224502-100G-KEA 127 EUR
SIALW224502-100G-K SIALW224502-1KG-K SIALW224502-5KG-K
5-Isopropyl-2-methylphenol, Sigma-Aldrich®
5-Isopropyl-2-methylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237
Melting Pt: 0.5
Density: 0.98 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00002236
CAS Number: 499-75-2
EINECS: 207-889-6
UN: 3265
ADR: 8 III