5-Isopropyl-2-methylphenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
282197-10G 282197-50G
SIAL282197-10GEA 62.1 EUR
SIAL282197-10G SIAL282197-50G
5-Isopropyl-2-methylphenol, Sigma-Aldrich®
5-Isopropyl-2-methylphenol
Formula: C₁₀H₁₄O
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 0.5 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00002236
CAS Number: 499-75-2
EINECS: 207-889-6
UN: 3265
ADR: 8,III