4,4'-Isopropylidenediphenol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Bisphenol A, BPA, 2,2-Bis(4-hydroxyphenyl)propane
M-Clarity™ quality level = MQ100
133027-500G 133027-2KG
SIAL133027-500GEA 37.3 EUR
SIAL133027-500G SIAL133027-2KG
4,4'-Isopropylidenediphenol, Sigma-Aldrich®
4,4'-Isopropylidenediphenol
Formula: C₁₅H₁₆O₂
MW: 228.29 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 155 °C
Density: 1.205 g/cm³ (20 °C)
Flash Pt: 227 °C
MDL Number: MFCD00002366
CAS Number: 80-05-7
EINECS: 201-245-8
Merck Index: 13,01296