2-(Bromomethyl)phenylboronic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-(Bromomethyl)benzeneboronic acid, Dihydroxy(2-(bromomethyl)phenyl)borane, (2-(Bromomethyl)phenyl)metaboric acid, 2-Bromomethylphenylboronic acid

M-Clarity™ quality level = MQ100

679453-1G 679453-5G
SIAL679453-1GEA 31.7 EUR
SIAL679453-1G SIAL679453-5G
2-(Bromomethyl)phenylboronic acid, Sigma-Aldrich®
2-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
MW: 214.85 g/mol
MDL Number: MFCD01318941
CAS Number: 91983-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR