(±)-β-Citronellol, Sigma-Aldrich®
Supplier: Merck
Synonyms: DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol
M-Clarity™ quality level = MQ100
C83201-100G C83201-5G
SIALC83201-100GEA 57.9 EUR
SIALC83201-100G SIALC83201-5G
(±)-β-Citronellol, Sigma-Aldrich®
(±)-β-Citronellol
Formula: C₁₀H₂₀O
Boiling Pt: 222 °C (1013 hPa)
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00002935
CAS Number: 106-22-9
EINECS: 203-375-0
UN: 3082
ADR: 9,III
Merck Index: 13,02354