2-Bromonicotinic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Bromo-3-pyridinecarboxylic acid, 2-Bromopyridine-3-carboxylic acid

M-Clarity™ quality level = MQ100

632465-1G 632465-5G
SIAL632465-1GEA 26.1 EUR
SIAL632465-1G SIAL632465-5G
2-Bromonicotinic acid, Sigma-Aldrich®
2-Bromonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
MDL Number: MFCD01646068
CAS Number: 35905-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR