Citral, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,7-Dimethylocta-2,6-dienal
M-Clarity™ quality level = MQ300
W230308-1KG-K W230308-20KG-K W230308-9KG-K
SIALW230308-1KG-KEA 99.9 EUR
SIALW230308-1KG-K SIALW230308-20KG-K SIALW230308-9KG-K
Citral, Sigma-Aldrich®
Citral
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 229
Melting Pt: < –10 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 98
Storage Temperature: Ambient
MDL Number: MFCD00006997
CAS Number: 5392-40-5
EINECS: 226-394-6
Merck Index: 13,02346