Citral, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3,7-Dimethylocta-2,6-dienal
M-Clarity™ quality level = MQ100
W230316-1KG-K
W230316-9KG-K
SIALW230316-1KG-KEA
204
EUR
SIALW230316-1KG-K
SIALW230316-9KG-K
Citral, Sigma-Aldrich®
Citral
Formula:
C₁₀H₁₆O MW: 152.24 g/mol Boiling Pt: 229 Melting Pt: < –10 °C Density: 0.89 g/cm³ (20 °C) Flash Pt: 98 Storage Temperature: Ambient |
MDL Number:
MFCD00006997 CAS Number: 5392-40-5 EINECS: 226-394-6 Merck Index: 13,02346 |