Citral, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,7-Dimethylocta-2,6-dienal
M-Clarity™ quality level = MQ100
C83007-100ML C83007-500ML C83007-5ML
SIALC83007-100MLEA 65.2 EUR
SIALC83007-100ML SIALC83007-500ML SIALC83007-5ML
Citral, Sigma-Aldrich®
Citral
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: < –10 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00006997
CAS Number: 5392-40-5
EINECS: 226-394-6
Merck Index: 13,02346