3-Aminophenylboronic acid monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Dihydroxy(3-aminophenyl)borane monohydrate, 3-Aminobenzeneboronic acid monohydrate, 3-Aminobenzeneboronic acid, 3-Aminophenylboronic Acid Monohydrate, (3-Aminophenyl)metaboric acid monohydrate
M-Clarity™ quality level = MQ200
287512-1G
287512-5G
SIAL287512-1GEA
40.3
EUR
SIAL287512-1G
SIAL287512-5G
3-Aminophenylboronic acid monohydrate, Sigma-Aldrich®
3-Aminophenylboronic acid monohydrate
Formula:
C₆H₈BNO₂·1H₂O MW: 154.96 g/mol |
MDL Number:
MFCD00149554 CAS Number: 206658-89-1 EINECS: 250-189-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...