Piperonyl alcohol, Sigma-Aldrich®

Supplier: Merck

Synonyms: Benzo[d][1,3]dioxol-5-ylmethanol, 3,4-(Methylenedioxy)benzyl alcohol, 1,3-Benzodioxol-5-ylmethanol

M-Clarity™ quality level = MQ100

P49406-100G P49406-25G
SIALP49406-100GEA 128 EUR
SIALP49406-100G SIALP49406-25G
Piperonyl alcohol, Sigma-Aldrich®
Piperonyl alcohol
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 51…54 °C
MDL Number: MFCD00005836
CAS Number: 495-76-1
EINECS: 207-808-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR