α-Angelicalactone, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Methylfuran-2(3H)-one
M-Clarity™ quality level = MQ300
W329304-100G-K W329304-1KG-K W329304-250G-K
SIALW329304-100G-KEA 207 EUR
SIALW329304-100G-K SIALW329304-1KG-K SIALW329304-250G-K
α-Angelicalactone, Sigma-Aldrich®
α-Angelicalactone
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.0904 g/cm³ (12 °C)
MDL Number: MFCD00005375
CAS Number: 591-12-8
EINECS: 209-701-8
Merck Index: 12,00686