α-Angelicalactone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5-Methylfuran-2(3H)-one
M-Clarity™ quality level = MQ100
A86406-100G
A86406-10G
SIALA86406-100GEA
191
EUR
SIALA86406-100G
SIALA86406-10G
α-Angelicalactone, Sigma-Aldrich®
α-Angelicalactone
Formula:
C₅H₆O₂ MW: 98.1 g/mol Boiling Pt: 249 °C (1013 hPa) Melting Pt: 0 °C Density: 1.0904 g/cm³ (12 °C) |
MDL Number:
MFCD00005375 CAS Number: 591-12-8 EINECS: 209-701-8 Merck Index: 12,00686 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...