Piperidin-4-ol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
128775-100G 128775-25G 128775-5G
SIAL128775-100GEA 185 EUR
SIAL128775-100G SIAL128775-25G SIAL128775-5G
Piperidin-4-ol, Sigma-Aldrich®
Piperidin-4-ol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 86…89 °C
Flash Pt: <100 °C
MDL Number: MFCD00005999
CAS Number: 5382-16-1
EINECS: 226-373-1
ADR: 8,III