(±)-γ-Octalactone, Sigma-Aldrich®
Supplier: Merck
Synonyms: (±)-Octan-4-olide, 5-Butyldihydrofuran-2(3H)-one
M-Clarity™ quality level = MQ300
W279617-100G-K W279617-1KG-K
SIALW279617-100G-KEA 183 EUR
SIALW279617-100G-K SIALW279617-1KG-K
(±)-γ-Octalactone, Sigma-Aldrich®
(±)-γ-Octalactone
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 234 °C (1013 hPa)
Density: 0.991 g/cm³ (0 °C)
MDL Number: MFCD00005402
CAS Number: 104-50-7
EINECS: 203-208-1