4-Phenylpiperidine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

639869-1G 639869-5G
SIAL639869-1GEA 49.2 EUR
SIAL639869-1G SIAL639869-5G
4-Phenylpiperidine, Sigma-Aldrich®
4-Phenylpiperidine
Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Boiling Pt: 257 °C (1013 hPa)
Melting Pt: 58…62 °C
Density: 0.967 g/cm³ (20 °C)
MDL Number: MFCD00006002
CAS Number: 771-99-3
EINECS: 212-243-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR