Ethyl-6-bromohexanoate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 6-Bromocaproic acid ethyl ester, 6-Bromohexanoic acid ethyl ester, Ethyl 6-bromohexanoate

M-Clarity™ quality level = MQ200

324728-100G 324728-25G
SIAL324728-100GEA 205 EUR
SIAL324728-100G SIAL324728-25G
Ethyl-6-bromohexanoate, Sigma-Aldrich®
Ethyl 6-bromohexanoate
Formula: BrH₂C(CH₂)₄COOCH₂CH₃
MW: 223.11 g/mol
Boiling Pt: 69…72 °C (0.1 mmHg)
Melting Pt: 33 °C
Density: 1.2592 g/cm³ (20 °C)
MDL Number: MFCD00000270
CAS Number: 25542-62-5
EINECS: 247-085-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR