ε-Hexalactone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
ε-Caprolactone, Oxepan-2-one, Hexan-6-olide
M-Clarity™ quality level = MQ200
704067-100G
704067-500G
SIAL704067-100GEA
45.4
EUR
SIAL704067-100G
SIAL704067-500G
ε-Hexalactone, Sigma-Aldrich®
ε-Hexalactone
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 225 °C (1013 hPa) Melting Pt: –1 °C Density: 1.0705 g/cm³ (20 °C) |
MDL Number:
MFCD00003267 CAS Number: 502-44-3 EINECS: 207-938-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...