(±)-Isonipecotic acid
Supplier: Thermo Scientific
Synonyms:
Piperidine-4-carboxylic acid, (±)-4-Piperidinecarboxylic acid, 4-Piperidinecarboxylic acid, Isonipecotic acid
SB00736.1
SB00736.10
SB00736.50
MAYBSB00736.1EA
0
EUR
MAYBSB00736.1
MAYBSB00736.10
MAYBSB00736.50
(±)-Isonipecotic acid
(±)-Isonipecotic acid
Formula:
C₆H₁₁NO₂ Boiling Pt: 264 °C (1013 hPa) Melting Pt: 157 °C |
MDL Number:
MFCD00006004 CAS Number: 498-94-2 EINECS: 207-872-3 Merck Index: 13,05208 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...