Boron trichloride 1 mol/l in heptane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
348325-100ML 348325-800ML
SIAL348325-100MLEA 66 EUR
SIAL348325-100ML SIAL348325-800ML
Boron trichloride 1 mol/l in heptane, Sigma-Aldrich®
Boron trichloride
Formula: BCl₃
Storage Temperature: Ambient
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 3384
ADR: 6.1,I