Chloroacetyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
22880-100ML 22880-1L 22880-500ML
SIAL22880-100MLEA 38.3 EUR
SIAL22880-100ML SIAL22880-1L SIAL22880-500ML
Chloroacetyl chloride, Sigma-Aldrich®
Chloroacetyl chloride
Formula: C₂H₂Cl₂O
MW: 112.94 g/mol
Boiling Pt: 105 °C (1013 hPa)
Melting Pt: –22.5 °C
Density: 1.426 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00000725
CAS Number: 79-04-9
EINECS: 201-171-6
UN: 1752
ADR: 6.1,I
Merck Index: 13,02074