Glycolic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Hydroxy acid, 2-Hydroxyacetic acid
M-Clarity™ quality level = MQ200
G8284-100G G8284-10MG G8284-25G G8284-500G
SIALG8284-100GEA 368 EUR
SIALG8284-100G SIALG8284-10MG SIALG8284-25G SIALG8284-500G
Glycolic acid, Sigma-Aldrich®
Glycolic acid
Formula: C₂H₄O₃
MW: 76.05 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 79…80 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II
Merck Index: 13,04511