Glycolic acid ≥70% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Hydroxy acid, 2-Hydroxyacetic acid
M-Clarity™ quality level = MQ100
420581-500ML 420581-100ML
SIAL420581-500MLEA 79.4 EUR
SIAL420581-500ML SIAL420581-100ML
Glycolic acid ≥70% in aqueous solution, Sigma-Aldrich®
Glycolic acid
Formula: C₂H₄O₃
MW: 76.05 g/mol
Density: 1.262 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3265
ADR: 8,II