Polyethylene glycol 1,000, Sigma-Aldrich®
Supplier: Merck
P3515-1KG P3515-250G P3515-500G
SIALP3515-1KGEA 62.1 EUR
SIALP3515-1KG SIALP3515-250G SIALP3515-500G
Polyethylene glycol 1,000, Sigma-Aldrich®
Polyethylene glycol 1,000
Formula: H(OCH₂CH₂)nOH
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 33…40 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 13,07651