Polyethylene glycol 10,000, Sigma-Aldrich®
Supplier: Merck
Synonyms: PEG 10000, Ethylene glycol polymer 10000, PEG 10,000
M-Clarity™ quality level = MQ200
P6667-1KG P6667-2KG P6667-500G
SIALP6667-1KGEA 76.6 EUR
SIALP6667-1KG SIALP6667-2KG SIALP6667-500G
Polyethylene glycol 10,000, Sigma-Aldrich®
Polyethylene glycol 10,000
Formula: H(OCH₂CH₂)nOH
MW: 10000 g/mol
Melting Pt: 63
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 265
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2