Polyethylene glycol 400, Sigma-Aldrich®
Supplier: Merck
P3265-1KG
P3265-500G
SIALP3265-1KGEA
85.6
EUR
SIALP3265-1KG
SIALP3265-500G
Polyethylene glycol 400, Sigma-Aldrich®
Polyethylene glycol 400
Formula:
H(OCH₂CH₂)nOH Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 4…8 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |