Titanium(IV) chloride 1 mol/l in toluene, AcroSeal®
Supplier: Thermo Scientific
Danger
Synonyms: Titanium tetrachloride
378161000 378168000
378161000.EA 76.2 EUR
378161000. 378168000.
Titanium(IV) chloride 1 mol/l in toluene, AcroSeal®
Titanium (IV) chloride
Formula: Cl₄Ti
MW: 189.69 g/mol
Density: 0.964 g/cm³
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00011267
CAS Number: 7550-45-0
EINECS: 231-441-9
UN: 3286
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear orange Solution
Titration Argentometric 0.95 to 1.1 M