Bacillol® Wipes
Supplier: BODE CHEMIE
9819110 9819110_1
BODE9819110EA 44.5 EUR
BODE9819110 BODE9819110_1
Bacillol® Wipes
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Alcohol-based rapid disinfectant wipes with comprehensive effectiveness for use in the Bacillol® wipes dispenser.
  • Ready-to-use disinfectant wipes soaked in Bacillol® AF for use in the Bacillol® Wipes dispenser
  • Free of dyes, perfumes and aldehydes
  • ECARF-certified: professional allergy- and asthmatic-friendly surface disinfectants
  • Can be used without gloves
  • Fast and broadly effective
  • Good wetting and fast drying
  • Tear-resistant and absorbent PET fleece
  • Wipes in dispenser can be used for 28 days
Can be used in the Bacillol® Wipes dispenser for quick disinfection of alcohol-resistant surfaces and medical inventory. In the healthcare sector, in laboratories as well as in commercial kitchens and in the food sector. In areas where a fast exposure and drying time is required.