Upright microscopes, Axio Lab.A1
Supplier: ZEISS
4909500003000 4909500006000 4300379010000 4909500002000
630-1271EA 0 EUR
630-1271 630-1272 630-2143 630-2078
Upright microscopes, Axio Lab.A1
Microscopes
The Axio Lab.A1 has been designed for biomedical applications.
  • All applications (brightfield, phase contrast, polarisation or fluorescence) can be performed with IC²S objectives
  • LED is available (optional)
  • Fluorescence LED comes with UV and blue excitations
  • Study small structures in a relaxed posture
  • Anti-fungal treatment for tropical zones
They offer flexibility in the analysis of defects, quality control and material testing. Stand versions for orthoscopy or conoscopy to examine bi-refringent materials. Please contact your local Avantor sales office for further details.
Certifikace: TÜV approved ergonomic design for high comfort.