VWR® Pharma-65 Tubing, Silicone
Supplier: Avantor
VWR®
VWRU89198-890EA 505.96 EUR
VWRU89198-890
VWR® Pharma-65 Tubing, Silicone
Tubing Transfer Tubing
Pharmaceutical grade silicone tubing for transferring of ultra-pure liquids, air or steam in pharmaceutical and biotechnological manufacturing processes.
  • Excellent flexibility and low extractables
  • Easily sterilised
  • High resiliency
  • Complete traceability
  • No impaired taste or odor
Certifikace: Meets or exceeds 21 CFR 177.2600, GMPs, USP Class VI and 3A Sanitary Standard procedures and European Pharmacopoeia Monograph 3.1.9. silicone elastomer for closures and tubing.
Balení: Standard package includes 15-meter coil, double bagged polyethylene bags in a cardboard box.