VWR® EndoGrade Glass Test Tubes
Supplier: Avantor
VWRU101076-598EA 980 EUR
VWRU101076-598
VWR® EndoGrade Glass Test Tubes
Tubes Test Tubes
Borosilicate glass test tubes with aluminium screw cap.
  • Capacity: 5 ml
  • Height: 50 mm
112 endotoxin-free (<0,005 EU/ml).