D-Alanine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
M-Clarity™ quality level = MQ200
A7377-100G
A7377-25G
A7377-5G
SIALA7377-100GEA
617
EUR
SIALA7377-100G
SIALA7377-25G
SIALA7377-5G
D-Alanine, Sigma-Aldrich®
D-Alanine
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 189 Melting Pt: 79 |
MDL Number:
MFCD00008077 CAS Number: 338-69-2 EINECS: 206-418-1 Merck Index: 13,00203 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...