4-Fluoro-3-nitrobenzaldehyde

Supplier: Thermo Scientific

SB02056DA SB02056-1
MAYBSB02056DAEA 0 EUR
MAYBSB02056DA MAYBSB02056-1
4-Fluoro-3-nitrobenzaldehyde
4-Fluoro-3-nitrobenzaldehyde
Formula: C₇H₄FNO₃
MW: 169.11 g/mol
MDL Number: MFCD01861388
CAS Number: 42564-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR