L-Alanine, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
M-Clarity™ quality level = MQ300
W381829-100G W381829-1KG
SIALW381829-100GEA 85.5 EUR
SIALW381829-100G SIALW381829-1KG
L-Alanine, Sigma-Aldrich®
L-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189
Melting Pt: 79
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203