L-Alanine, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
M-Clarity™ quality level = MQ200
A7469-100G A7469-10MG A7469-1KG A7469-25G
SIALA7469-100GEA 169 EUR
SIALA7469-100G SIALA7469-10MG SIALA7469-1KG SIALA7469-25G
L-Alanine, Sigma-Aldrich®
L-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189
Melting Pt: 79
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203