(4-Nitrophenyl)acetonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-Nitrobenzyl cyanide
M-Clarity™ quality level = MQ100
151572-100G
151572-25G
SIAL151572-100GEA
93.7
EUR
SIAL151572-100G
SIAL151572-25G
(4-Nitrophenyl)acetonitrile, Sigma-Aldrich®
(4-Nitrophenyl)acetonitrile
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Boiling Pt: 336 °C (1013 hPa) Melting Pt: 108…113 °C |
MDL Number:
MFCD00007372 CAS Number: 555-21-5 EINECS: 209-085-0 UN: 3439 ADR: 6.1,III Merck Index: 12,06688 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...