Propylene oxide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
(±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
82320-1L
82320-250ML
SIAL82320-1LEA
84.5
EUR
SIAL82320-1L
SIAL82320-250ML
Propylene oxide, Sigma-Aldrich®
Propylene oxide
Formula:
C₃H₆O MW: 58.08 g/mol Boiling Pt: 34.3 °C (1013 hPa) Melting Pt: –112 °C Density: 0.83 g/cm³ (20 °C) Flash Pt: –37 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005126 CAS Number: 75-56-9 EINECS: 200-879-2 UN: 1280 ADR: 3,I Merck Index: 12,08041 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...